missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enocyanin, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Enocyanin is an anthocyanin extracted from grapes. Enocyanin shows inhibitory effect on the leucine aminopeptidase, acid phosphatase, γ-glutamyl transpeptidase and esterase activity.
Specifications
Specifications
| Chemical Name or Material | Enocyanin |
| CAS | 11029-12-2 |
| Color | Brown |
| Purity Grade Notes | Research |
| Solubility Information | DMSO : ≥ 50 mg/mL ∣H2O : 5 mg/mL (Need ultrasonic) |
| SMILES | [Enocyanin] |
| Quantity | 100 mg |
| Grade | Research |
| Physical Form | Solid |
| For Use With (Application) | Neuroscience-Neuromodulation |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction