missing translation for 'onlineSavingsMsg'
Learn More

Enocitabine, TRC

Product Code. 30383685
Change view
Click to view available options
Quantity:
5 mg
100 mg
250 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383685 5 mg -
30374028 250 mg -
30376433 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383685 Supplier Toronto Research Chemicals Supplier No. TRCE5575005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55726-47-1
Molecular Formula C31 H55 N3 O6
Molecular Weight (g/mol) 565.78
IUPAC Name N-[1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]docosanamide
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C31H55N3O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-27(36)32-26-22-23-34(31(39)33-26)30-29(38)28(37)25(24-35)40-30/h22-23,25,28-30,35,37-38H,2-21,24H2,1H3,(H,32,33,36,39)/t25-,28-,29+,30-/m1/s1
Formula Weight 565.4091
Chemical Name or Material Enocitabine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.