missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16421-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| ENO3 | |
| Polyclonal | |
| Unconjugated | |
| ENO3 | |
| Beta enolase, ENO3, Enolase 3, enolase 3 (beta, muscle), MSE, Muscle specific enolase, Skeletal muscle enolase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13808, 2027, 25438 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13929, P15429, P21550 | |
| ENO3 | |
| ENO3 Fusion Protein Ag9613 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction