Learn More
Invitrogen™ ENO3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120444
Description
Positive test controls include: Mouse heart, Mouse skeletal muscle, Rat heart, Rat skeletal muscle. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MAMQKIFARE ILDSRGNPTV EVDLHTAKGR FRAAVPSGAS TGIYEALELR DGDKGRYLGK GVLKAVENIN NTLGPALLQK KLSVVDQEKV DKFMIELDGT ENKSKFGANA ILGVSLAVCK AGAAEKGVPL YRHIADLAGN PDLILPVPAF NVINGGSHAG NKLAMQEFMI LPVGASSFKE AMRIGAEVYH.
This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| ENO3 | |
| Polyclonal | |
| Unconjugated | |
| ENO3 | |
| 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; BBE; beta beta enolase; beta-enolase; cb883; ENO1; ENO3; Eno-3; eno3 protein; enob; enolase 1, (alpha); enolase 3; enolase 3 (beta, muscle); enolase 3, (beta, muscle); enolase 3, beta muscle; enolase 3, beta, muscle; fj24f12; GSD13; hypothetical protein LOC540303; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); muscle-specific enolase; Skeletal muscle enolase; wu:fj24f12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13808, 2027, 25438 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13929, P15429, P21550 | |
| ENO3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human ENO3 (NP_001967.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.