missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENO3 Antibody (008), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90275AF488
This item is not returnable.
View return policy
Description
ENO3 Monoclonal specifically detects ENO3 in Human samples. It is validated for ELISA.
Specifications
| ENO3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 8 | |
| ELISA | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, beta-enolase, EC 4.2.1, EC 4.2.1.11, Enolase 3, enolase 3 (beta, muscle), enolase 3, (beta, muscle), GSD13, MSE, Muscle-specific enolase, Skeletal muscle enolase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ENO3 (AAH17249.1; Met1-Lys434). | |
| 0.1 mL | |
| metabolism | |
| 2027 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction