missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENO1 Rabbit anti-Human, Mouse, Rat, CoraLite 595, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL595-11204-100UL
This item is not returnable.
View return policy
Description
ENO1 Polyclonal antibody apecifically detetects ENO1 antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.Specifications
| ENO1 | |
| Polyclonal | |
| CoraLite 595 | |
| BC015641 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Flow Cytometry | |
| 1000 μg/mL | |
| PBS with 0.09% sodium azide. | |
| ENO1 | |
| ENO1 fusion protein Ag1692 | |
| 100 μL | |
| 2023 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction