missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENO1 Monoclonal Antibody (GT186)

Description
Keep as concentrated solution. Predicted reactivity: Mouse (94%), Rat (93%), Xenopus laevis (90%), Chicken (93%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (93%), Guinea pig (95%). Positive Control: 293T, A549, H1299, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Enolase (2-phosphogly-cerate hydrolyase or phosphopyruvate hydrates) is a glycolytic enzyme that catalyzes the dehydration and conversion of 2-phosphoglycerate to phosphoenolpyruvate. It comprises three distinct subunits, alpha, beta and gamma. Non Neuronal Enolase(ENO1) is an isoform of mammalian enolase and is composed of 2 alpha subunits. The gene for ENO1 also encodes a shorter monomeric structural lens protein, tau-crystallin. Multifunctional enzyme that, as well as its role in glycolysis, plays a part in various processes such as growth control, hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its ability to serve as a receptor and activator of plasminogen on the cell surface of several cell-types such as leukocytes and neurons. Stimulates immunoglobulin production.Used as a diagnostic marker for many tumors and, in the heterodimeric form, alpha/gamma, as a marker for hypoxic brain injury after cardiac arrest. Also marker for endometriosis. Antibodies against alpha-enolase are present in sera from patients with cancer-associated retinopathy syndrome (CAR), a progressive blinding disease which occurs in the presence of systemic tumor growth, primarily small-cell carcinoma of the lung and other malignancies. Is identified as an autoantigen in Hashimoto encephalopathy (HE) a rare autoimmune disease associated with Hashimoto thyroiditis (HT). HT is a disorder in which destructive processes overcome the potential capacity of thyroid replacement leading to hypothyroidism.
Specifications
Specifications
| Antigen | ENO1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | GT186 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | ENO1 |
| Gene Accession No. | P04764, P06733 |
| Gene Alias | 0610008I15; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydro-lyase; AL022784; alpha enolase; alpha enolase like 1; alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; c-Myc promoter binding protein; C-myc promoter-binding protein; Eno1; Eno-1; ENO1L1; enoa; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; enolase-alpha; epididymis secretory protein Li 17; HEL-S-17; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?