missing translation for 'onlineSavingsMsg'
Learn More

Enfuvirtide Acetate (>90%), TRC

Product Code. 30380194
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380194 10 mg
30383155 50 mg
30377732 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380194 Supplier Toronto Research Chemicals Supplier No. TRCE55560010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914454-00-5
Molecular Formula C196H294N50O61
Molecular Weight (g/mol) 4356.13
Synonym Enfuvirtide acetate, L-Phenylalaninamide, N-acetyl-L-tyrosyl-L-threonyl-L-seryl-L-leucyl-L-isoleucyl-L-histidyl-L-seryl-L-leucyl-L-isoleucyl-L-α-glutamyl-L-α-glutamyl-L-seryl-L-glutaminyl-L-asparaginyl-L-glutaminyl-L-glutaminyl-L-α-glutamyl-L-lysyl-L-asparaginyl-L-α-glutamyl-L-glutaminyl-L-α-glutamyl-L-leucyl-L-leucyl-L-α-glutamyl-L-leucyl-L-α-aspartyl-L-lysyl-L-tryptophyl-L-alanyl-L-seryl-L-leucyl-L-tryptophyl-L-asparaginyl-L-tryptophyl-, acetate, N-Acetyl-L-tyrosyl-L-threonyl-L-seryl-L-leucyl-L-isoleucyl-L-histidyl-L-seryl-L-leucyl-L-isoleucyl-L-α-glutamyl-L-α-glutamyl-L-seryl-L-glutaminyl-L-asparaginyl-L-glutaminyl-L-glutaminyl-L-α-glutamyl-L-lysyl-L-asparaginyl-L-α-glutamyl-L-glutaminyl-L-α-glutamyl-L-leucyl-L-leucyl-L-α-glutamyl-L-leucyl-L-α-aspartyl-L-lysyl-L-tryptophyl-L-alanyl-L-seryl-L-leucyl-L-tryptophyl-L-asparaginyl-L-tryptophyl-L-phenylalaninamide acetate
SMILES CC(O)=O.CN[C@@H]([C@H](O)C)C(N[C@@H](CO)C(N[C@@H](CC(C)C)C(N[C@H](C(N[C@H](C(N[C@@H](CO)C(N[C@@H](CC(C)C)C(N[C@H](C(N[C@@H](CCC(O)=O)C(N[C@@H](CCC(O)=O)C(N[C@@H](CO)C(N[C@@H](CCC(N)=O)C(C([C@H](CC(N)=O)NN[C@@H](CCC(N)=O)C(N[C@@H](CCC(N)=O)C(N[C@@H](CCC(O)=O)C(N[C@@H](CCCCN)C(N[C@@H](cc([nH])=O)C(N[C@@H](CCC(O)=O)C(N[C@@H](CCC(N)=O)C(N[C@@H](CCC(O)=O)C(N[C@@H](CC(C)C)C(N[C@@H](CC(C)C)C(N[C@@H](CCC(O)=O)C(N[C@@H](CC(C)C)C(C([C@@H](NN[C@@H](CCCCN)C(N[C@H](C(N[C@@H](C(N[C@@H](CO)C(N[C@@H](CC(C)C)C(N[C@H](C(N[C@@H](CC(N)=O)C(N[C@H](C(N[C@H](C(N)=O)CC1=CC=CC=C1)#O)CC2=CNC3=CC=CC=C32)=O)=O)CC4=CNC5=CC=CC=C54)=O)=O)=O)C)=O)CC6=CNC7=CC=CC=C76)=O)CC(O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)[C@@H](C)CC)=O)=O)=O)CC8=CNC=N8)=O)[C@@H](C)CC)=O)=O)=O

Specifications

Quantity 10 mg
Formula Weight 4358.8
Chemical Name or Material Enfuvirtide acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.