missing translation for 'onlineSavingsMsg'
Learn More

Endoxifen (E-isomer), MedChemExpress

Product Code. p-200098406
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480078

Brand: Medchem Express HY18719D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Endoxifen E-isomer (E-Endoxifen), an E-isomer of Endoxifen, is an impurity in Endoxifen Z-isomer drug substance. Endoxifen E-isomer exhibits antiestrogenic effects.

Endoxifen E-isomer (E-Endoxifen), an E-isomer of Endoxifen, is an impurity in Endoxifen Z-isomer drug substance. Endoxifen E-isomer exhibits antiestrogenic effects.

Specifications

Chemical Name or Material Endoxifen (E-isomer)
CAS 114828-90-9
Color White
Purity Grade Notes Research
Molecular Formula C25H27NO2
Quantity 5 mg
Synonym E-Endoxifen
Solubility Information DMSO : ≥ 32 mg/mL (85.68 mM)
SMILES OC1=CC=C(/C(C2=CC=C(OCCNC)C=C2)=C(C3=CC=CC=C3)/CC)C=C1
Molecular Weight (g/mol) 373.49
Formula Weight 373.49
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.