missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoxifen E-isomer hydrochloride, MedChemExpress
Description
Endoxifen E-isomer hydrochloride (E-Endoxifen hydrochloride), an E-isomer of Endoxifen, is an impurity in Endoxifen Z-isomer drug substance. Endoxifen E-isomer hydrochloride exhibits antiestrogenic effects.
Specifications
Specifications
| Chemical Name or Material | Endoxifen E-isomer hydrochloride |
| CAS | 1197194-61-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H28ClNO2 |
| Quantity | 5 mg |
| Synonym | E-Endoxifen hydrochloride |
| Solubility Information | DMSO : ≥ 30 mg/mL (73.18 mM) |
| SMILES | OC1=CC=C(/C(C2=CC=C(OCCNC)C=C2)=C(C3=CC=CC=C3)/CC)C=C1.[H]Cl.[E] |
| Molecular Weight (g/mol) | 409.95 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?