missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endov Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570136
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 77%.
ENDOV gene ontology annotations related to this gene include DNA repair.
Specifications
| Endov | |
| Polyclonal | |
| Unconjugated | |
| ENDOV | |
| A730011L01Rik; Endonuclease V; Endov; hEndoV; inosine-specific endoribonuclease; novel endonuclease V protein; putative endonuclease FLJ39025 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 338371 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8C9A2 | |
| ENDOV | |
| synthetic peptide directed towards the middle region of human Endov (aa 138-187). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction