missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Endotoxin-Free Buffers for NucleoBond™ Xtra EF Kits
Click to view available options
Quantity:
600 mL
900 mL
1000 mL
3000 mL
Chemical Name or Material:
ELU-EF
ENDO-EF
EQU-EF
FIL-EF
LYS-EF
NEU-EF
WASH-EF
Unit Size:
1000mL
3000mL
600mL
900mL
Description
The new generation of anion exchanger – ultra-fast, endotoxin-free plasmid DNA with high yield.
- Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Midi prep in approximately 45 min, Maxi prep in 50 min
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 400 μg (Midi) and 1000 μg (Maxi)
- NucleoBond™ Xtra Plus EF kits contain NucleoBond Finalizers to speed up DNA precipitation
Specifications
Specifications
| Chemical Name or Material | ELU-EF |
| CAS | 64-17-5d |
| Color | Colorless |
| For Use With (Application) | Endotoxin-free plasmid DNA purification |
| For Use With (Equipment) | NucleoBond™ Xtra EF Kit |
| pH | 8.5 to 9.5 |
| Physical Form | Liquid |
| Quantity | 900 mL |
| Formulation | 2-Propanol, Potassium chloride, Chemicals mixture <1%, Water |
| Odor | Alcoholic |
Safety and Handling
Product Identifier
- Buffer ELU-EF (900 mL)
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : 2-propanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction