missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Endotoxin-Free Buffers for NucleoBond™ Xtra EF Kits

Product Code. 15810908
Click to view available options
Quantity:
600 mL
900 mL
1000 mL
3000 mL
Chemical Name or Material:
ELU-EF
ENDO-EF
EQU-EF
FIL-EF
LYS-EF
NEU-EF
WASH-EF
Unit Size:
1000mL
3000mL
600mL
900mL
This item is not returnable. View return policy

Product Code. 15810908

Brand: Macherey-Nagel™ 740393.3000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supplemental buffer solutions for use with NucleoBond™ Xtra EF kits for ultra fast, endotoxin-free purification of plasmid DNA with high yield.

The new generation of anion exchanger – ultra-fast, endotoxin-free plasmid DNA with high yield.

  • Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design – Midi prep in approximately 45 min, Maxi prep in 50 min
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 400 μg (Midi) and 1000 μg (Maxi)
  • NucleoBond™ Xtra Plus EF kits contain NucleoBond Finalizers to speed up DNA precipitation

Specifications

Chemical Name or Material ELU-EF
CAS 64-17-5d
Color Colorless
For Use With (Application) Endotoxin-free plasmid DNA purification
For Use With (Equipment) NucleoBond™ Xtra EF Kit
pH 8.5 to 9.5
Physical Form Liquid
Quantity 3000 mL
Formulation 2-Propanol, Potassium chloride, Chemicals mixture <1%, Water
Odor Alcoholic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.