missing translation for 'onlineSavingsMsg'
Learn More

Endothion, TRC

Product Code. 30136039
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136039 100 mg 100mg
30134672 50 mg 50mg
30040535 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136039 Supplier Toronto Research Chemicals Supplier No. TRCE555210100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2778-04-3
Molecular Formula C9 H13 O6 P S
Molecular Weight (g/mol) 280.235
Synonym Phosphorothioic acid, O,O-dimethyl ester, S-ester with 2-(mercaptomethyl)-5-methoxy-4H-pyran-4-one (6CI,7CI,8CI), 5-Methoxy-2-(dimethoxyphosphinylthiomethyl)pyrone-4, AC-18737, Endocid, Endocide, Endothion, Niagara 5767, S-(5-Methoxy-4-pyron-2-ylmethyl) O,O-dimethyl phosphorothioate, S-[(5-Methoxy-4-oxo-4H-pyran-2-yl)methyl] O,O-dimethylphosphorothioate, Phosphorothioic acid, S-[(5-methoxy-4-oxo-4H-pyran-2-yl)methyl] O,O-dimethyl ester, S-((5-Methoxy-4-oxo-4H-pyran-2-yl)methyl) O,O-dimethyl Phosphorothioate;
IUPAC Name 2-(dimethoxyphosphorylsulfanylmethyl)-5-methoxy-pyran-4-one
SMILES COC1=COC(=CC1=O)CSP(=O)(OC)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13O6PS/c1-12-9-5-15-7(4-8(9)10)6-17-16(11,13-2)14-3/h4-5H,6H2,1-3H3
Formula Weight 280.017
Chemical Name or Material Endothion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.