missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endothelium Monoclonal Antibody (MRC OX43)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA190752
This item is not returnable.
View return policy
Description
MA1-90752 detects Endothelium from rat samples.
The endothelium is the layer of thin specialized epithelium, comprising a simple squamous layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall (simple squamous epithelium). Endothelial cells line the entire circulatory system, from the heart to the smallest capillary. These cells reduce friction of the flow of blood.
Specifications
| Endothelium | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| MRC OX43 | |
| Unconjugated | |
| Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells | |
| Rat peritoneal macrophages. | |
| 500 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction