Learn More
Invitrogen™ Endothelin B Receptor Recombinant Rabbit Monoclonal Antibody (JM74-10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532630
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications
| Endothelin B Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P24530 | |
| EDNRB | |
| Synthetic peptide within Human Endothelin B Receptor aa 58-91. | |
| 100 μL | |
| Primary | |
| Human, Zebrafish | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| JM74-10 | |
| Unconjugated | |
| EDNRB | |
| ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1910 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.