missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin A Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530589
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG, GL261. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.
Specifications
| Endothelin A Receptor | |
| Polyclonal | |
| Unconjugated | |
| EDNRA | |
| Ednra; Endor; Endothelin A; endothelin A receptor; endothelin receptor subtype A; endothelin receptor type A; endothelin type A receptor; endothelin-1 receptor; endothelin-1-specific receptor; ETa; ET-A; ETAR; ET-AR; ETA-R; ETRA; G protein coupled receptor; G protein-coupled receptor; Gpcr10; G-protein coupled receptor 10; hET-AR; MFDA; RGD1559432 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13617, 1909, 24326, 373908 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.78 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P25101, P26684, Q61614 | |
| EDNRA | |
| Synthetic peptide corresponding to a region within amino acids 363 and 427 of Human Endothelin A Receptor | |
| 100 μL | |
| Primary | |
| Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction