missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Endothelin A Receptor Polyclonal Antibody
GREENER_CHOICE

Product Code. 11375723
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375723 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11375723 Supplier Invitrogen™ Supplier No. PA3065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA3-065 detects Endothelin Receptor A from human, mouse, and rat samples. PA3-065 has been successfully used in Western blot, immunocytochemistry, and immunohistochemistry procedures. By western blot, this antibody detects a 46kDa protein corresponding to mouse Endothelin Receptor A.

This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin A Receptor
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene EDNRA
Gene Accession No. P25101, P26684, Q61614
Gene Alias Ednra; Endor; Endothelin A; endothelin A receptor; endothelin receptor subtype A; endothelin receptor type A; endothelin type A receptor; endothelin-1 receptor; endothelin-1-specific receptor; ETa; ET-A; ETAR; ET-AR; ETA-R; ETRA; G protein coupled receptor; G protein-coupled receptor; Gpcr10; G-protein coupled receptor 10; hET-AR; MFDA; RGD1559432
Gene Symbols EDNRA
Host Species Rabbit
Immunogen COOH-terminal amino acids of mouse ETA (408-427) KNQEQNNHNTERSSHKDSMN coupled to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13617, 1909, 24326
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.