missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endothelin A Receptor Monoclonal Antibody (485711)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561682
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.
Specifications
| Endothelin A Receptor | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P25101 | |
| EDNRA | |
| NS0 mouse myeloma cell line transfected with human EDRNA. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| 485711 | |
| Unconjugated | |
| EDNRA | |
| Ednra; Endor; Endothelin A; endothelin A receptor; endothelin receptor subtype A; endothelin receptor type A; endothelin type A receptor; endothelin-1 receptor; endothelin-1-specific receptor; ETa; ET-A; ETAR; ET-AR; ETA-R; ETRA; G protein coupled receptor; G protein-coupled receptor; Gpcr10; G-protein coupled receptor 10; hET-AR; MFDA; RGD1559432 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1909 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction