missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin-1 Antibody (TR.ET.48.5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-526AF700
This item is not returnable.
View return policy
Description
Endothelin-1 Monoclonal specifically detects Endothelin-1 in Human, Mouse, Rat, Porcine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Endothelin-1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| P05305 | |
| EDN1 | |
| Pure endothelin-1 conjugated to KLH. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1906 | |
| Human, Mouse, Rat, Pig, Canine, Rabbit, Sheep | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| TR.ET.48.5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Not yet tested in other species. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction