missing translation for 'onlineSavingsMsg'
Learn More

Endothelin-1 Antibody (3G10) [PerCP], Novus Biologicals™

Product Code. 30737013 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737013 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737013 Supplier Novus Biologicals Supplier No. FAB3440C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Endothelin-1 Monoclonal antibody specifically detects Endothelin-1 in Human samples. It is validated for ELISA,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin-1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3G10
Conjugate PerCP
Formulation PBS
Gene Alias endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1
Host Species Rat
Immunogen Human Endothelin-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1906
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.