missing translation for 'onlineSavingsMsg'
Learn More

Endothelin-1 Antibody (2E1) [PE], Novus Biologicals™

Product Code. 30735286 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735286 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735286 Supplier Novus Biologicals Supplier No. FAB34401P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Endothelin-1 Monoclonal antibody specifically detects Endothelin-1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin-1
Applications ELISA
Classification Monoclonal
Clone 20
Conjugate PE
Formulation PBS
Gene Alias endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1
Host Species Mouse
Immunogen Human Endothelin-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1906
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.