missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelial Lipase Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-118UV
This item is not returnable.
View return policy
Description
Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Endothelial Lipase | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719 | |
| Rabbit | |
| 0.1 mL | |
| 9388 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LIPG | |
| An N-terminal synthetic peptide made to human Endothelial Lipase [UniProtNo. Q9Y5X9] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction