missing translation for 'onlineSavingsMsg'
Learn More

Endothelial Lipase Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18258164 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18258164 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18258164 Supplier Novus Biologicals Supplier No. NB400111AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelial Lipase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Gene Symbols LIPG
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to human Endothelial Lipase [UniProtNo. Q9Y5X9]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9388
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.