missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Endothelial Cell Growth Medium-2 Bullet Kit

Product Code. 11645440
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11645440 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11645440 Supplier Lonza™ Supplier No. CC3162
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Endothelial Cell Growth Medium-2 Bulletkit is part of a collection of tools optimized to grow certain cell types.

Media products are supplied in several formats:
  • Basal Medium -– contains no growth factors, cytokines, or supplements
  • SingleQuots™ Kit – growth factors, cytokines, and supplements
  • BulletKit™ – basal medium and SingleQuots™ Kit packaged together
The following Endothelial Cell Growth Medium BulletKits™ are available:
  • EGM™-2 BulletKit™ – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
  • EGM™ BulletKit™ – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
  • New! EGM™-Plus BulletKit™ – Enhanced version of EGM™ Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM™ Medium.
  • EGM™-2MV BulletKit™ – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
  • EGM™ -MV BulletKit™ – Bovine Microvascular Cells, contains 5% FBS and BBE. Does not contain exogenous VEGF.

Order Info

dimProductType

TRUSTED_SUSTAINABILITY

Specifications

Product Type Growth Medium
Quantity 1 Kit
Content And Storage No BBE (bovine brain extract), hEGF, hydrocortisone, GA-1000 (Gentamicin, Amphotericin-B), FBS (fetal bovine serum) 10 ml, VEGF, hFGF-B, R3-IGF-1, Ascorbic Acid, Heparin
compliance-icons compliance-icons compliance-icons
Product Identifier
  • EGM-2 Endothelial SingleQuots Kit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol, Gentamicin,sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.