missing translation for 'onlineSavingsMsg'
Learn More

Endosulfan Sulfate, TRC

Product Code. 30374119
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374119 100 mg
30372844 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374119 Supplier Toronto Research Chemicals Supplier No. TRCE555500100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031-07-8
Molecular Formula C9 H6 Cl6 O4 S
Molecular Weight (g/mol) 422.92
Synonym 6,9-Methano-2,4,3-benzodioxathiepin, 6,7,8,9,10,10-hexachloro-1,5,5a,6,9,9a-hexahydro-, 3,3-dioxide, 5-Norbornene-2,3-dimethanol, 1,4,5,6,7,7-hexachloro-, cyclic sulfate (7CI,8CI), 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dimethanol cyclic sulfate, 6,7,8,9,10,10-Hexachloro-1,5,5a,6,9,9a-hexahydro-6,9-methano-2,4,3-benzodioxathiepin 3,3-dioxide, 6,7,8,9,10,10-Hexachloro-1,5,5a,6,9,9a-hexahydro-6,9-methano-2,4,3-benzodioxathiepin 3,6-dioxide, Benzoepin sulfate, Endosulfan III, Endosulfan cyclic sulfate, Endosulfan sulfate, Endosulfate, Thiodan sulfate
SMILES ClC1=C(Cl)[C@@]2(Cl)C3COS(=O)(=O)OCC3[C@]1(Cl)C2(Cl)Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H6Cl6O4S/c10-5-6(11)8(13)4-2-19-20(16,17)18-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3?,4?,7-,8+
Formula Weight 419.8118
Chemical Name or Material Endosulfan-sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.