missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Endostatin Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHCOL18A1X5

Additional Details : Weight : 0.33820kg

 View more versions of this product

Product Code. 17175399

  • £2320.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

The Human Endostatin ELISA quantitates Hu Endostatin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Endostatin. Principle of the method The Human Endostatin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

COL18A1 is the alpha chain of type XVIII collagen. This collagen is one of the multiplexins, extracellular matrix proteins that contain multiple triple-helix domains (collagenous domains) interrupted by non-collagenous domains. The proteolytically produced C-terminal fragment of type XVIII collagen is endostatin, a potent antiangiogenic protein. Mutations in this gene are associated with Knobloch syndrome. The main features of this syndrome involve retinal abnormalities, so type XVIII collagen may play an important role in retinal structure and in neural tube closure.
Specifications

Specifications

P39060
10 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
80781
<12%
Wash Buffer, Stop Solution, Chromogen, Pre-coated 96 well plates, Standard, Assay Diluent Concentrate, Biotinylated Detection Antibody, Adhesive Plate Covers, SAV-HRP
5 x 96 Tests
Angiogenesis
2°C to 8°C
Human
4 hr. 45 min.
13.7 pg/mL to 10,000 pg/mL
10 pg/mL to 10,000 pg/mL
Biotin
Other Proteins
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
KNO,KNO1,KS
<10%
HRP
RUO
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L
KNO, KNO1, KS
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers