missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endosialin/CD248/TEM1 Antibody (OTI4B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46129
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
Endosialin/CD248/TEM1 Monoclonal specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Western Blot.
Specifications
| Endosialin/CD248/TEM1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1 | |
| Mouse | |
| 80.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| CD248 | |
| Human recombinant protein fragment corresponding to amino acids 20-300 of human CD248(NP_065137) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 57124 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction