missing translation for 'onlineSavingsMsg'
Learn More

Endosialin/CD248/TEM1 Antibody (2A7), Novus Biologicals™

Product Code. 18418379 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418379 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418379 Supplier Novus Biologicals Supplier No. H00057124M12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Endosialin/CD248/TEM1 Monoclonal antibody specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endosialin/CD248/TEM1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2A7
Conjugate Unconjugated
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_065137
Gene Alias 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1
Host Species Mouse
Immunogen CD248 (NP_065137.1, 18 a.a. ∽ 757 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QDPWAAEPRAACGPSSCYALFPRRRTFLEAWRACRELGGDLATPRTPEEAQRVDSLVGAGPASRLLWIGLQRQARQCQLQRPLRGFTWTTGDQDTAFTNWAQPASGGPCPAQRCVALEASGEHRWLEGSCTLAVDGYLCQFGFEGACPALQDEAGQAGPAVYTTPFHLVSTEFEWLPFGSVAAVQCQAGRGASLLCVKQPEGGVGWSRAGPLCLGTGCSPDNGGCEHECVEEVDGHVSCRCTEGFRLAADGRSCEDPCAQAPCEQQCEPGGPQGYSCHCRLGFRPAEDDPHRCVDTDECQIAGVCQQMCVNYVGGFECYCSEGHELEADGISCSPAGAMGAQASQDLGDELLDDGEDEEDEDEAWKAFNGGWTEMPGILWMEPTQPPDFALAYRPSFPEDREPQIPYPEPTWPPPLSAPRVPYHSSVLSVTRPVVVSATHPTLPSAHQPPVIPATHPALSRDHQIPVIAANYPDLPSAYQPGILSVSHSAQPPAHQPPMISTKYPELFPAHQSPMFPDTRVAGTQTTTHLPGIPPNHAPLVTTLGAQLPPQAPDALVLRTQATQLPIIPTAQPSLTTTSRSPVSPAHQISVPAATQPAALPTLLPSQSPTNQTSPISPTHPHSKAPQIPREDGPSPKLALWLPSPAPTAAPTALGEAGLAEHSQRDDRWLLVALLVPTCVFLVVLLALGIVYCTRCGPHAPNKRITDCYRWVIHAGSKSPTEPMPPRGSLTGVQTCRTSV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57124
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.