missing translation for 'onlineSavingsMsg'
Learn More

Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18372594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372594 100 μg 100µL
18341824 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18372594 Supplier Novus Biologicals Supplier No. NBP317840100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Polyclonal antibody specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Endorepellin/Perlecan/Heparan Sulfate Proteoglycan
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RGMVFGIPDGVLELVPQRGPCPDGHFYLEHSAACLPCFCFGITSVCQSTRRFRDQIRLRFDQPDDFKGVNVTMPAQPGTPPLSSTQLQIDPSL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 3339
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.