missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endophilin B1/Bif-1 Antibody (30A882.1.1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-24733CL1
This item is not returnable.
View return policy
Description
Endophilin B1/Bif-1 Monoclonal antibody specifically detects Endophilin B1/Bif-1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Simple Western,Immunocytochemistry/Immunofluorescence
Specifications
| Endophilin B1/Bif-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Bax-interacting factor 1, Bif-1dJ612B15.2, CGI-61, endophilin-B1, KIAA0491endophilin B1, SH3 domain-containing GRB2-like protein B1, SH3-containing protein SH3GLB1, SH3-domain GRB2-like endophilin B1, SH3-domain, GRB2-like, endophilin B1 | |
| A full-length human Bif-1 recombinant protein was used as immunogen. | |
| 0.1 mL | |
| Apoptosis, Mitochondrial Mediated Pathway | |
| 51100 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Simple Western | |
| 30A882.1.1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction