missing translation for 'onlineSavingsMsg'
Learn More

Endophilin A1/SH3GL2 Antibody [PE], Novus Biologicals™

Product Code. 30738410 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738410 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738410 Supplier Novus Biologicals Supplier No. AF8006P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Endophilin A1/SH3GL2 Polyclonal antibody specifically detects Endophilin A1/SH3GL2 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endophilin A1/SH3GL2
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias CNSA2FLJ25015, EEN-B1bA335L15.1 (SH3-domain GRB2-like 2), Endophilin-1, endophilin-A1, FLJ20276, SH3 domain protein 2A, SH3 domain-containing GRB2-like protein 2, SH3D2AEndophilin A1 BAR domain, SH3-domain GRB2-like 2, SH3P4endophilin-1
Host Species Sheep
Immunogen E. coli-derived recombinant human Endophilin A1/SH3GL2, Ala217-Pro293, Accession # Q99962
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6456
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.