missing translation for 'onlineSavingsMsg'
Learn More

Endophilin A1/SH3GL2 Antibody (2G6), Novus Biologicals™

Product Code. 18368979 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368979 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368979 Supplier Novus Biologicals Supplier No. H00006456M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Endophilin A1/SH3GL2 Monoclonal antibody specifically detects Endophilin A1/SH3GL2 in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endophilin A1/SH3GL2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2G6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003017
Gene Alias CNSA2FLJ25015, EEN-B1bA335L15.1 (SH3-domain GRB2-like 2), Endophilin-1, endophilin-A1, FLJ20276, SH3 domain protein 2A, SH3 domain-containing GRB2-like protein 2, SH3D2AEndophilin A1 BAR domain, SH3-domain GRB2-like 2, SH3P4endophilin-1
Host Species Mouse
Immunogen SH3GL2 (NP_003017, 64 a.a. ~ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SRAKLSMINTMSKIRGQEKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMREL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6456
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.