missing translation for 'onlineSavingsMsg'
Learn More

Endophenazine A, TRC

Product Code. 30384059
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384059 1 mg
30369459 25 mg
30369335 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384059 Supplier Toronto Research Chemicals Supplier No. TRCE5554801MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86125-71-5
Molecular Formula C18H16N2O2
Molecular Weight (g/mol) 292.33
Synonym 9-(3-Methyl-2-buten-1-yl)-1-phenazinecarboxylic Acid, 9-(3-Methyl-2-butenyl)-1-phenazinecarboxylic Acid
IUPAC Name 9-(3-methylbut-2-enyl)phenazine-1-carboxylic acid
SMILES CC(=CCc1cccc2nc3cccc(C(=O)O)c3nc12)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H16N2O2/c1-11(2)9-10-12-5-3-7-14-16(12)20-17-13(18(21)22)6-4-8-15(17)19-14/h3-9H,10H2,1-2H3,(H,21,22)
Formula Weight 292.1212
Chemical Name or Material Endophenazine A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.