missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endomucin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547648
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant human Endomucin-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.
Specifications
| Endomucin | |
| Polyclonal | |
| Unconjugated | |
| Emcn | |
| 0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 59308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9R0H2 | |
| Emcn | |
| E. coli-derived recombinant mouse Endomucin Glu21-Glu90. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction