missing translation for 'onlineSavingsMsg'
Learn More

Endomucin/Mucin -14 Antibody [Biotin], Novus Biologicals™

Product Code. 30734407 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734407 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734407 Supplier Novus Biologicals Supplier No. AF7206B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Endomucin/Mucin -14 Polyclonal antibody specifically detects Endomucin/Mucin -14 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endomucin/Mucin -14
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias endomucin, MUC-14
Host Species Goat
Immunogen E. coli-derived recombinant human Endomucin, Asn19-Gly139, Accession # Q9ULC0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51705
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.