missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Endomorphin-2

Product Code. 15448529
Change view
Click to view available options
Quantity:
5mg
25mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448529 25mg 25mg
15438529 5mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448529 Supplier Thermo Scientific Chemicals Supplier No. J66953.MB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141801-26-5

Specifications

Color White
Content And Storage Store at -20°C
Format Solid
Molecular Weight (g/mol) 571.678
Product Type Endomorphin-2
Quantity 25mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.