missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglycan/PODXL2 Antibody (211816), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1524CL1
This item is not returnable.
View return policy
Description
Endoglycan/PODXL2 Monoclonal antibody specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| Endoglycan/PODXL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2 | |
| Mouse myeloma cell line NS0-derived recombinant human Endoglycan/PODXL2, Gly33-Thr500, Accession # Q9NZ53 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 211816 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 50512 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction