missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Antibody (MJ7/18), Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-77666
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface).
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Flow (Cell Surface) | |
| ENG | |
| Inflamed mouse skin | |
| 0.5 mg | |
| Cancer, Cell Biology, Endothelial Cell Markers, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2022 | |
| Mouse | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| MJ7/18 | |
| Unconjugated | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| CD105 (MJ7/18). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction