missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Antibody (MEM-226), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human, Rat samples. It is validated for Flow Cytometry, Flow (Cell Surface)
Specifications
Specifications
| Antigen | Endoglin/CD105 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-226 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry, Flow (Cell Surface) |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 |
| Host Species | Mouse |
| Immunogen | Recombinant full length protein (Human). Expressed in vaccinia virus containing CD105 cDNA. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?