missing translation for 'onlineSavingsMsg'
Learn More

Endoglin/CD105 Antibody (MEM-226), DyLight 350, Novus Biologicals™

Product Code. 18400535 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400535 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400535 Supplier Novus Biologicals Supplier No. NB500452UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Cell Surface), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endoglin/CD105
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Flow Cytometry (Cell Surface), CyTOF
Classification Monoclonal
Clone MEM-226
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Host Species Mouse
Immunogen Recombinant full length protein (Human). Expressed in vaccinia virus containing CD105 cDNA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.