missing translation for 'onlineSavingsMsg'
Learn More

Endoglin/CD105 Antibody (ENG/1327), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54517PE

Product Code. 18640857

  • £481.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endoglin/CD105
Monoclonal
PE
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Recombinant human Endoglin/CD105 protein fragment (aa74-251) (exact sequence is proprietary) (Uniprot: P17813)
100 μg
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
2022
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunofluorescence, CyTOF
ENG/1327
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.