missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Antibody (ENG/1326), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £409.00
Specifications
| Antigen | Endoglin/CD105 |
|---|---|
| Clone | ENG/1326 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287280
|
Novus Biologicals
NBP2-53327-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284610
|
Novus Biologicals
NBP2-53327-100UG |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| Endoglin/CD105 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 2022 | |
| Recombinant human Endoglin protein fragment (aa74-251) (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| ENG/1326 | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
| ENG | |
| IgG1 | |
| Protein A or G purified | |
| 84 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title