missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENDOGL1 Recombinant Rabbit Monoclonal Antibody (K01_1K79)
GREENER_CHOICE

Product Code. 30827702
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827702 100 μL
30826729 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827702 Supplier Invitrogen™ Supplier No. MA563269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ENDOGL1 Recombinant Monoclonal Antibody for Western Blot

This gene encodes an endo/exonuclease with 5'-3' exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5' end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENDOGL1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1K79
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene EXOG
Gene Accession No. Q9Y2C4
Gene Alias AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein
Gene Symbols EXOG
Host Species Rabbit
Immunogen A synthetic peptide of human ENDOGL1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9941
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.