missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENDOGL1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143251
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_005098.2; NP_001138936.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes an endo/exonuclease with 5'-3' exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5' end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.
Specifications
| ENDOGL1 | |
| Polyclonal | |
| Unconjugated | |
| EXOG | |
| AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 9941 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9Y2C4 | |
| EXOG | |
| Peptide with sequence C-KAKEQSGTQIRKP (aa 355-367). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction