missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOGL1 Antibody (S04-9F5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19513-100ul
This item is not returnable.
View return policy
Description
ENDOGL1 Monoclonal antibody specifically detects ENDOGL1 in Human samples. It is validated for Western Blot
Specifications
| ENDOGL1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S04-9F5 | |
| Unconjugated | |
| EC 3.1.30, EC 3.1.30.-, Endo G-like 1, endo/exonuclease (5'-3'), endonuclease G-like, ENDOGL1mitochondrial, ENDOGL2, Endonuclease G-like 1MGC125945, ENGLA, ENGLB, ENGLMGC125944 | |
| A synthetic peptide of human ENDOGL1 (Uniprot # Q9Y2C4) | |
| 100 μg | |
| Endocrinology | |
| 9941 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction