missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOGL1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33188-100ul
This item is not returnable.
View return policy
Description
ENDOGL1 Monoclonal antibody specifically detects ENDOGL1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| ENDOGL1 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| EC 3.1.30, EC 3.1.30.-, Endo G-like 1, endo/exonuclease (5'-3'), endonuclease G-like, ENDOGL1mitochondrial, ENDOGL2, Endonuclease G-like 1MGC125945, ENGLA, ENGLB, ENGLMGC125944 | |
| A synthetic peptide corresponding to a sequence within amino acids 269-368 of human ENDOGL1 (Q9Y2C4).,, Sequence:, LQDLEKLSGLVFFPHLDRTSDIRNICSVDTCKLLDFQEFTLYLSTRKIEGARSVLRLEKIMENLKNAEIEPDDYFMSRYEKKLEELKAKEQSGTQIRKPS | |
| 100 μL | |
| Endocrinology | |
| 9941 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction