missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endocan Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144096
This item is not returnable.
View return policy
Description
Endocan Polyclonal Antibody for IHC (P), ELISA
Endothelial cell-specific molecule 1 is a protein that in humans is encoded by the ESM1 gene. This gene encodes a secreted protein which is mainly expressed in the endothelial cells in human lung and kidney tissues. The expression of this gene is regulated by cytokines, suggesting that it may play a role in endothelium-dependent pathological disorders. The transcript contains multiple polyadenylation and mRNA instability signals. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Endocan | |
| Polyclonal | |
| Unconjugated | |
| ESM1 | |
| 0610042H23Rik; AV004503; endocan; endothelial cell specific molecule 1; endothelial cell-specific molecule 1; ESM1; ESM-1; ESM-1 secretory protein; IGFBP-rp6; PG25; pineal specific PG25 protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11082 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9NQ30 | |
| ESM1 | |
| Recombinant Human Endothelial cell-specific molecule 1 protein (20-184AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction