Learn More
Invitrogen™ Endocan (ESM1) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMESM1
Code : NEW
Additional Details : Weight : 0.00400kg
Description
The Mouse Endocan (Ms ESM1) ELISA quantitates Ms Endocan in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Endocan. Principle of the method The Mouse Endocan solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes a secreted protein which is mainly expressed in the endothelial cells in human lung and kidney tissues. The expression of this gene is regulated by cytokines, suggesting that it may play a role in endothelium-dependent pathological disorders. The transcript contains multiple polyadenylation and mRNA instability signals. Two transcript variants encoding different isoforms have been found for this gene.Specifications
Q9QYY7 | |
0.1 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
71690 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Development | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
0.123-30 ng/mL | |
0.1ng/mL to 30ng/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA antibody pair detects mouse Endocan, it shows no significant cross-reactivity with recombinant human Endocan. | |
Mouse | |
Colorimetric Microplate Reader | |
0610042H23Rik,AV004503,ESM-1 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
0610042H23Rik, AV004503, ESM-1 | |
1 hr. 20 min. |