missing translation for 'onlineSavingsMsg'
Learn More

Endocan/ESM-1 Antibody (RM0079-5J44), DyLight 594, Novus Biologicals™

Product Code. 18423604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423604 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423604 Supplier Novus Biologicals Supplier No. NBP121710DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Endocan/ESM-1 Monoclonal antibody specifically detects Endocan/ESM-1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endocan/ESM-1
Applications Western Blot
Classification Monoclonal
Clone RM0079-5J44
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias endocan, endothelial cell-specific molecule 1, ESM-1
Host Species Rat
Immunogen Mouse recombinant Endocan
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11082
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.